siRNA Details
| Gene Symbol: Olfr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207664.2 | NP_997547.1 | 1 | 231 | ||
| GenBank mRNA | AY245537.1 | - | 1 | 274 | ||
| BC138117.1 | - | 1 | 231 | 8861136 | ||
| BC138118.1 | - | 1 | 231 | 8861137 | ||
| Gene Symbol: Olfr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030553.2 | NP_085030.2 | 1 | 154 | ||
| GenBank mRNA | BC130240.1 | - | 1 | 155 | 40142140 | |

Olfr151
Genomic Map