siRNA Details
| Gene Symbol: Calm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020036.4 | NP_064420.2 | 1 | 718 | ||
| GenBank mRNA | AK003648.1 | - | 1 | 719 | ||
| AK009664.1 | - | 1 | 720 | |||
| AK009956.1 | - | 1 | 719 | |||
| AY293058.1 | - | 1 | 701 | |||
| BC060284.1 | - | 1 | 704 | 30293362 | ||

Calm4
Genomic Map