siRNA Details
| Gene Symbol: Klb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031180.2 | NP_112457.1 | 1 | 644 | ||
| GenBank mRNA | AF165170.1 | - | 1 | 644 | ||
| AF165171.1 | - | 1 | 648 | |||
| AF178429.1 | - | 1 | 639 | |||
| AK133823.1 | - | 1 | 670 | |||
| BC138008.1 | - | 1 | 733 | 8861027 | ||
| BC138010.1 | - | 1 | 733 | 8861029 | ||
| KC810035.1 | - | 1 | 647 | |||

Klb
Genomic Map