siRNA Details
| Gene Symbol: Lmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053098.2 | NP_444328.1 | 1, 2 | 389 | ||
| GenBank mRNA | AK147141.1 | - | 1, 2 | 390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmod2| Gene Symbol: Lmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053098.2 | NP_444328.1 | 1, 2 | 389 | ||
| GenBank mRNA | AK147141.1 | - | 1, 2 | 390 | ||