siRNA Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080560.3 | NP_542127.1 | 4 | 1375 | ||
| GenBank mRNA | AK005302.1 | - | 4 | 1393 | ||
| AK150930.1 | - | 1 | 316 | |||
| AK161968.1 | - | 4 | 1391 | |||
| BC067069.1 | - | 4 | 1376 | 30550024 | ||
| BC132630.1 | - | 4 | 1350 | 40130907 | ||
| BC138211.1 | - | 4 | 1350 | 8861230 | ||

Ube2n
Genomic Map