siRNA Details
| Gene Symbol: Pcdhb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053132.3 | NP_444362.3 | 1 | 1204 | ||
| GenBank mRNA | AF326300.1 | - | 1 | 1000 | ||
| AK029340.1 | - | 1 | 1183 | |||
| AK034205.1 | - | 1 | 1206 | |||
| AK043979.1 | - | 1 | 1163 | |||
| AK046463.1 | - | 1 | 1187 | |||
| AY013789.1 | - | 1 | 1000 | |||

Pcdhb7
Genomic Map