siRNA Details
| Gene Symbol: Pcdhb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053133.1 | NP_444363.1 | 1 | 1261 | ||
| GenBank mRNA | AY013790.1 | - | 1 | 1261 | ||
| BC116268.1 | - | 1 | 1262 | |||
| BC116269.1 | - | 1 | 1254 | |||
| BC127160.1 | - | 1 | 1261 | 40094058 | ||

Pcdhb8
Genomic Map