siRNA Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053161.2 | NP_444391.1 | 4 | 534 | ||
| GenBank mRNA | AK143575.1 | - | 4 | 563 | ||
| BC030074.1 | - | 4 | 534 | 1514322 | ||
| BC096568.1 | - | 4 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl27| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053161.2 | NP_444391.1 | 4 | 534 | ||
| GenBank mRNA | AK143575.1 | - | 4 | 563 | ||
| BC030074.1 | - | 4 | 534 | 1514322 | ||
| BC096568.1 | - | 4 | 541 | |||