siRNA Details
| Gene Symbol: Tmem40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168256.1 | NP_001161728.1 | 3 | 316 | ||
| NM_001168257.1 | NP_001161729.1 | 3 | 316 | |||
| NM_144805.2 | NP_659054.2 | 3 | 184 | |||
| GenBank mRNA | BC019416.1 | - | 3 | 146 | 5151159 | |
| BY104106.1 | - | 3 | 171 | |||
| BY279100.1 | - | 3 | 316 | |||
| CT010303.1 | - | 2 | 114 | |||
| CV561368.1 | - | 3 | 258 | |||
| CX564997.1 | - | 3 | 215 | |||

Tmem40
Genomic Map