siRNA Details
| Gene Symbol: Mfsd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133829.2 | NP_598590.2 | 4 | 2136 | ||
| NM_178081.4 | NP_835182.1 | 4 | 2136 | |||
| GenBank mRNA | AB247936.1 | - | 3 | 1751 | ||
| AK035995.1 | - | 4 | 2137 | |||
| AK046866.1 | - | 4 | 2280 | |||
| BC017534.1 | - | 3 | 1245 | 4488390 | ||
| BC029324.1 | - | 3 | 1493 | |||

Mfsd6
Genomic Map