siRNA Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006417.4 | NP_006408.3 | 5 | 827 | ||
| NR_135640.1 | 4 | 363 | ||||
| NR_135641.1 | 5 | 830 | ||||
| XM_005270380.3 | XP_005270437.1 | 5 | 2065 | |||
| XM_011540516.2 | XP_011538818.1 | 5 | 2065 | |||
| XM_017000083.1 | XP_016855572.1 | 5 | 2065 | |||
| GenBank mRNA | AK302527.1 | - | 5 | 788 | ||
| AK316278.1 | - | 4 | 320 | |||
| AK316476.1 | - | 5 | 785 | |||
| BC022870.1 | - | 5 | 792 | 4071404 | ||

IFI44
Genomic Map