siRNA Details
Gene Symbol: SLU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006425.4 | NP_006416.3 | 16 | 1824 | ||
XM_005265793.3 | XP_005265850.1 | 16 | 2171 | |||
XM_011534418.2 | XP_011532720.1 | 17 | 1874 | |||
GenBank mRNA | AF101074.1 | - | 15 | 1669 | ||
AK024875.1 | - | 16 | 1744 | |||
AK314564.1 | - | 16 | 1744 | |||
BC010634.1 | - | 16 | 1750 | 3871882 |
