siRNA Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006426.2 | NP_006417.2 | 11 | 1333 | ||
| XM_005252658.3 | XP_005252715.1 | 11 | 1322 | |||
| XM_011539125.2 | XP_011537427.1 | 12 | 1336 | |||
| XM_017015482.1 | XP_016870971.1 | 11 | 2597 | |||
| XM_017015483.1 | XP_016870972.1 | 12 | 2750 | |||
| GenBank mRNA | AB006713.1 | - | 11 | 1327 | ||
| BC029280.1 | - | 11 | 1217 | |||
| BC136329.1 | - | 11 | 1202 | 9020316 | ||
| BC136330.1 | - | 11 | 1202 | 9020317 | ||
| Y10976.1 | - | 11 | 1210 | |||

DPYSL4
Genomic Map