siRNA Details
| Gene Symbol: SIVA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006427.3 | NP_006418.2 | 4 | 738 | ||
| NM_021709.2 | NP_068355.1 | 3 | 543 | |||
| XM_011536360.2 | XP_011534662.1 | 4 | 1660 | |||
| GenBank mRNA | AK124078.1 | - | 1473 | |||
| AK128704.1 | - | 2 | 4118 | |||
| AK223332.1 | - | 4 | 724 | |||
| BC034562.1 | - | 3 | 517 | 3860364 | ||

SIVA1
Genomic Map