siRNA Details
| Gene Symbol: TBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006453.2 | NP_006444.2 | 14 | 1470 | ||
| GenBank mRNA | AK095076.1 | - | 6 | 1646 | ||
| BC007620.1 | - | 6 | 654 | |||
| BC010231.1 | - | 14 | 1470 | 3455740 | ||
| BC014007.2 | - | 14 | 1442 | 4040312 | ||
| BC035409.1 | - | 14 | 1426 | 5165833 | ||
| U02609.1 | - | 14 | 1418 | |||
More Information
Set Membership: |
|

TBL3
Genomic Map