siRNA Details
| Gene Symbol: GAS2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278730.1 | NP_001265659.1 | 5 | 1150 | ||
| NM_006478.4 | NP_006469.2 | 5 | 1178 | |||
| NM_152236.2 | NP_689422.1 | 5 | 1183 | |||
| XM_011529824.1 | XP_011528126.1 | 6 | 1302 | |||
| XM_011529825.1 | XP_011528127.1 | 5 | 1111 | |||
| XM_017028533.1 | XP_016884022.1 | 5 | 1828 | |||
| GenBank mRNA | AK093386.1 | - | 5 | 1120 | ||
| AK122829.1 | - | 4 | 2020 | |||
| AK223602.1 | - | 5 | 1122 | |||
| AK225484.1 | - | 5 | 1122 | |||
| AK301715.1 | - | 4 | 605 | |||
| BC001782.1 | - | 5 | 1118 | 3355168 | ||
| BC007624.1 | - | 5 | 1118 | 3357387 | ||
| BC011047.1 | - | 5 | 1121 | 4178571 | ||
| Y07846.1 | - | 5 | 1104 | |||

GAS2L1
Genomic Map