siRNA Details
| Gene Symbol: IGF2BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006547.2 | NP_006538.2 | 10, 11 | 1468 | ||
| XM_006715639.2 | XP_006715702.1 | 9, 10 | 1096 | |||
| XM_011515089.2 | XP_011513391.1 | 11, 12 | 1781 | |||
| XM_011515090.2 | XP_011513392.1 | 7, 8 | 2347 | |||
| XM_011515091.2 | XP_011513393.1 | 8, 9 | 1413 | |||
| XM_011515092.2 | XP_011513394.1 | 11, 12 | 1154 | |||
| XM_011515093.2 | XP_011513395.1 | 10, 11 | 1026 | |||
| GenBank mRNA | AF117108.1 | - | 10, 11 | 1468 | ||
| BC065269.1 | - | 10, 11 | 1447 | 6154021 | ||
| BX640800.1 | - | 4, 5 | 479 | |||
| BX648488.1 | - | 3, 4 | 353 | |||
| U76705.1 | - | 10, 11 | 1452 | |||
| U97188.1 | - | 10, 11 | 1452 | |||

IGF2BP3
Genomic Map