siRNA Details
| Gene Symbol: MTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006635.2 | NP_001006636.1 | 6 | 707 | ||
| NM_001319097.1 | NP_001306026.1 | 5 | 503 | |||
| NM_001319098.1 | NP_001306027.1 | 5 | 503 | |||
| NM_006554.4 | NP_006545.1 | 5 | 503 | |||
| XM_011510497.2 | XP_011508799.1 | 6 | 600 | |||
| GenBank mRNA | AF053551.1 | - | 5 | 476 | ||
| AK001287.1 | - | 5 | 455 | |||
| AK289359.1 | - | 5 | 454 | |||
| AK303564.1 | - | 6 | 632 | |||
| AK315918.1 | - | 6 | 643 | |||
| BC017271.2 | - | 6 | 637 | 4635884 | ||
| BC067831.1 | - | 5 | 482 | 5298149 | ||
| BC088359.1 | - | 5 | 438 | 30378040 | ||
| BG699784.1 | - | 6 | 711 | |||
| BU165147.1 | - | 5 | 434 | |||
| CX165048.1 | - | 5 | 396 | |||

MTX2
Genomic Map