siRNA Details
| Gene Symbol: DMRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130865.2 | NP_001124337.1 | 6 | 1760 | ||
| NM_006557.6 | NP_006548.1 | 4 | 2321 | |||
| NM_181872.4 | NP_870987.2 | 4 | 1553 | |||
| XM_011517687.1 | XP_011515989.1 | 6 | 4101 | |||
| XM_011517690.2 | XP_011515992.1 | 5 | 2181 | |||
| XM_011517694.2 | XP_011515996.1 | 3 | 1712 | |||
| XM_017014213.1 | XP_016869702.1 | 6 | 2503 | |||
| XM_017014214.1 | XP_016869703.1 | 5 | 2178 | |||
| XM_017014215.1 | XP_016869704.1 | 4 | 2022 | |||
| GenBank mRNA | AF284223.1 | - | 3 | 1848 | ||
| AF284224.1 | - | 4 | 1431 | |||
| AF284225.1 | - | 3 | 1353 | |||
| AK290794.1 | - | 6 | 2066 | |||
| BC030204.1 | - | 6 | 1760 | |||
| BC136493.1 | - | 3 | 1432 | 9020482 | ||
| BX647565.1 | - | 5 | 1367 | |||

DMRT2
Genomic Map