siRNA Details
| Gene Symbol: CTCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006565.3 | NP_006556.1 | 3 | 722 | ||
| XM_005255775.3 | XP_005255832.1 | 3 | 722 | |||
| XM_017022868.1 | XP_016878357.1 | 3 | 472 | |||
| XM_017022869.1 | XP_016878358.1 | 3 | 471 | |||
| GenBank mRNA | AK314804.1 | - | 3 | 722 | ||
| BC014267.2 | - | 3 | 556 | 3840827 | ||
| BT009915.1 | - | 1 | 278 | |||
| U25435.1 | - | 3 | 569 | |||

CTCF
Genomic Map