siRNA Details
| Gene Symbol: CD226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303618.1 | NP_001290547.1 | 2 | 775 | ||
| NM_006566.3 | NP_006557.2 | 3 | 777 | |||
| XM_005266642.3 | XP_005266699.1 | 3 | 876 | |||
| XM_017025524.1 | XP_016881013.1 | 2 | 775 | |||
| XM_017025526.1 | XP_016881015.1 | 1 | 32 | |||
| GenBank mRNA | AJ537553.1 | - | 2 | 357 | ||
| AK313199.1 | - | 2 | 776 | |||
| BC074787.2 | - | 2 | 362 | 30915562 | ||
| GU935335.1 | - | 3 | 524 | |||
| U56102.1 | - | 3 | 536 | |||

CD226
Genomic Map