siRNA Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282425.1 | NP_001269354.1 | 2 | 323 | ||
| NM_006572.5 | NP_006563.2 | 2 | 633 | |||
| XM_011524202.1 | XP_011522504.1 | 2 | 427 | |||
| GenBank mRNA | AF493902.1 | - | 2 | 424 | ||
| AK301699.1 | - | 3 | 558 | |||
| AK302400.1 | - | 2 | 323 | |||
| AK313672.1 | - | 2 | 427 | |||
| BC036756.1 | - | 2 | 478 | 5548632 | ||
| L22075.1 | - | 2 | 465 | |||

GNA13
Genomic Map