siRNA Details
| Gene Symbol: PNMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007257.5 | NP_009188.1 | 3 | 939 | ||
| XM_011544365.2 | XP_011542667.1 | 3 | 1038 | |||
| GenBank mRNA | AB020690.1 | - | 1 | 346 | ||
| AF037365.1 | - | 1 | 164 | |||
| AK055325.1 | - | 3 | 881 | |||
| BC036489.1 | - | 3 | 869 | 5262276 | ||
| BC047515.1 | - | 3 | 844 | 5294654 | ||
| BC062301.1 | - | 1 | 420 | 6580976 | ||

PNMA2
Genomic Map