siRNA Details
| Gene Symbol: GMEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319674.1 | NP_001306603.1 | 5 | 554 | ||
| NM_006582.3 | NP_006573.2 | 5 | 584 | |||
| NM_024482.2 | NP_077808.1 | 5 | 551 | |||
| XM_011540518.2 | XP_011538820.1 | 5 | 687 | |||
| XM_011540519.2 | XP_011538821.1 | 5 | 527 | |||
| XM_011540521.2 | XP_011538823.1 | 5 | 656 | |||
| XM_011540522.2 | XP_011538824.1 | 5 | 496 | |||
| XM_017000087.1 | XP_016855576.1 | 5 | 702 | |||
| GenBank mRNA | AF099013.1 | - | 4 | 448 | ||
| AF173868.1 | - | 5 | 479 | |||
| AF203694.1 | - | 5 | 478 | |||
| AK000892.1 | - | 5 | 550 | |||
| AK023126.1 | - | 5 | 471 | |||
| AK315195.1 | - | 5 | 530 | |||
| AK315436.1 | - | 5 | 488 | |||
| BC001473.2 | - | 5 | 501 | 3138481 | ||

GMEB1
Genomic Map