siRNA Details
| Gene Symbol: BST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004335.3 | NP_004326.1 | 1 | 168 | ||
| GenBank mRNA | AK223124.1 | - | 1 | 106 | ||
| AK291099.1 | - | 1 | 113 | |||
| AK303593.1 | - | 1 | 138 | |||
| BC033873.1 | - | 1 | 168 | 5217945 | ||
| D28137.1 | - | 1 | 96 | |||

BST2
Genomic Map