siRNA Details
| Gene Symbol: SIX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016932.4 | NP_058628.3 | 2 | 1418 | ||
| XM_005264100.3 | XP_005264157.1 | 2 | 1666 | |||
| GenBank mRNA | AF136940.1 | - | 2 | 1177 | ||
| AF332196.1 | - | 2 | 1115 | |||
| AF332197.1 | - | 2 | 1418 | |||
| BC024033.2 | - | 2 | 1398 | 3027992 | ||

SIX2
Genomic Map