siRNA Details
| Gene Symbol: KLRA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028045.1 | 4 | 744 | |||
| GenBank mRNA | AF047445.1 | - | 4 | 474 | ||
| AK290077.1 | - | 3 | 445 | |||
| AK296103.1 | - | 4 | 481 | |||
| AK304677.1 | - | 3 | 440 | |||
| AY334570.1 | - | 2 | 288 | |||
| BC069672.1 | - | 4 | 447 | 7262588 | ||
| BC069697.1 | - | 4 | 447 | 7262600 | ||
| BC069734.1 | - | 4 | 447 | 7262612 | ||
| BC101471.1 | - | 4 | 461 | 8068977 | ||
| BC101473.1 | - | 4 | 464 | 8068979 | ||

KLRA1P
Genomic Map