siRNA Details
| Gene Symbol: TOB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016272.3 | NP_057356.1 | 2 | 806 | ||
| XM_005261315.2 | XP_005261372.1 | 2 | 2017 | |||
| XM_006724105.3 | XP_006724168.1 | 2 | 1323 | |||
| XM_017028539.1 | XP_016884028.1 | 2 | 3099 | |||
| GenBank mRNA | AB035207.1 | - | 1 | 313 | ||
| AB051450.1 | - | 2 | 565 | |||
| BC028919.1 | - | 2 | 555 | 4940977 | ||
| BC038957.1 | - | 2 | 579 | 5786640 | ||
| CR456594.1 | - | 2 | 259 | |||
| CR541684.1 | - | 1 | 99 | |||

TOB2
Genomic Map