siRNA Details
Gene Symbol: Gpr150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175495.2 | NP_780704.1 | 1 | 222 | ||
GenBank mRNA | AK047609.1 | - | 1 | 222 | ||
AY255541.1 | - | 1 | 26 | |||
BC118612.1 | - | 1 | 202 | 40107174 | ||
BC118614.1 | - | 1 | 203 | 40107172 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
