siRNA Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011815.5 | NP_035945.1 | 11 | 2093 | ||
| GenBank mRNA | AF061744.1 | - | 11 | 2069 | ||
| AK046759.1 | - | 2 | 1237 | |||
| AK172164.1 | - | 4 | 325 | |||
| BC141352.1 | - | 11 | 2034 | 9055964 | ||

Fyb
Genomic Map