siRNA Details
| Gene Symbol: Cdh24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199470.2 | NP_955764.1 | 8 | 1555 | ||
| GenBank mRNA | BC057373.1 | - | 1509 | 6817232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh24| Gene Symbol: Cdh24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199470.2 | NP_955764.1 | 8 | 1555 | ||
| GenBank mRNA | BC057373.1 | - | 1509 | 6817232 | ||