siRNA Details
| Gene Symbol: Lrrc52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013382.2 | NP_001013400.1 | 1 | 666 | ||
| GenBank mRNA | BC087947.1 | - | 1 | 666 | 30303788 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc52| Gene Symbol: Lrrc52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013382.2 | NP_001013400.1 | 1 | 666 | ||
| GenBank mRNA | BC087947.1 | - | 1 | 666 | 30303788 | |