siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 1775 | ||
| NM_001163797.1 | NP_001157269.1 | 6 | 1865 | |||
| NM_013844.2 | NP_038872.1 | 6 | 1871 | |||
| GenBank mRNA | AB024005.1 | - | 6 | 1855 | ||
| AK013868.1 | - | 6 | 1866 | |||
| AK044831.1 | - | 5 | 1745 | |||
| AK143158.1 | - | 1 | 271 | |||
| AK146092.1 | - | 6 | 1832 | |||
| AK153887.1 | - | 6 | 1807 | |||
| BC018171.1 | - | 6 | 1854 | 3989444 | ||

Zfp68
Genomic Map