siRNA Details
| Gene Symbol: Adam29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175939.3 | NP_787953.2 | 2 | 346 | ||
| GenBank mRNA | AY190759.1 | - | 2 | 330 | ||
| BB614467.1 | - | 2 | 348 | |||
| BC048676.1 | - | 2 | 344 | |||
| BC119089.1 | - | 2 | 327 | 8733838 | ||
| BC119091.1 | - | 2 | 327 | 8733840 | ||

Adam29
Genomic Map