siRNA Details
| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173432.2 | NP_775608.1 | 2 | 423 | ||
| GenBank mRNA | AK053397.1 | - | 2 | 457 | ||
| AK153861.1 | - | 2 | 470 | |||
| AK154570.1 | - | 2 | 453 | |||
| AK166338.1 | - | 2 | 446 | |||
| AK178313.1 | - | 1 | 364 | |||
| AK178805.1 | - | 1 | 364 | |||
| AK214424.1 | - | 1 | 364 | |||
| BC050128.1 | - | 2 | 423 | 30024345 | ||

Pskh1
Genomic Map