siRNA Details
| Gene Symbol: Olfr1000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011695.1 | NP_001011695.1 | 1 | 406 | ||
| Gene Symbol: Olfr996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146437.2 | NP_666648.2 | 1 | 432 | ||
| GenBank mRNA | BC120782.1 | - | 1 | 432 | 40129705 | |
| BC120784.1 | - | 1 | 432 | 40129707 | ||

Olfr1000
Genomic Map