siRNA Details
| Gene Symbol: Olfr429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146722.2 | NP_666933.2 | 1 | 590 | ||
| GenBank mRNA | BC127973.1 | - | 1 | 590 | 40135130 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr429| Gene Symbol: Olfr429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146722.2 | NP_666933.2 | 1 | 590 | ||
| GenBank mRNA | BC127973.1 | - | 1 | 590 | 40135130 | |