siRNA Details
| Gene Symbol: Greb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252071.1 | NP_001239000.1 | 29 | 5422 | ||
| NM_015764.4 | NP_056579.2 | 31 | 5828 | |||
| GenBank mRNA | AB093248.1 | - | 14 | 2756 | ||
| AF180470.1 | - | 30 | 5484 | |||
| AK017875.1 | - | 6 | 1045 | |||
| AK147337.1 | - | 29 | 5356 | |||
| BC160931.1 | - | 30 | 5354 | |||

Greb1
Genomic Map