siRNA Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080850.2 | NP_543126.2 | 10 | 2973 | ||
| GenBank mRNA | AK028435.1 | - | 10 | 2975 | ||
| AK083528.1 | - | 10 | 2944 | |||
| AK083590.1 | - | 10 | 2939 | |||
| AK129063.1 | - | 10 | 2863 | |||
| AK172031.1 | - | 10 | 2921 | |||
| BC150821.1 | - | 9 | 2685 | 9087732 | ||

Pask
Genomic Map