siRNA Details
| Gene Symbol: Lrrc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033371.3 | NP_001028543.1 | 11 | 1913 | ||
| NM_001170788.1 | NP_001164259.1 | 14 | 2242 | |||
| NM_001170789.1 | NP_001164260.1 | 11 | 1919 | |||
| GenBank mRNA | AK133042.1 | - | 11 | 1913 | ||
| BC036564.1 | - | 9 | 1530 | |||
| BC150957.1 | - | 11 | 1905 | 9087868 | ||
| BC172120.1 | - | 11 | 1911 | 9054406 | ||

Lrrc36
Genomic Map