siRNA Details
| Gene Symbol: Agbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178755.3 | NP_848870.2 | 17 | 2548 | ||
| GenBank mRNA | AK029744.1 | - | 17 | 2581 | ||
| AK029932.1 | - | 17 | 2549 | |||
| AK030138.1 | - | 17 | 2536 | |||
| AK030182.1 | - | 17 | 2595 | |||
| DQ867026.1 | - | 17 | 2581 | |||
| DQ867027.1 | - | 17 | 2581 | |||
| DQ867028.1 | - | 17 | 2581 | |||
| DQ867029.1 | - | 16 | 2473 | |||
| DQ867030.1 | - | 15 | 2419 | |||

Agbl2
Genomic Map