siRNA Details
| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181819.2 | NP_861540.2 | 1, 2 | 230 | ||
| GenBank mRNA | AY308804.1 | - | 1, 2 | 200 | ||
| BC113144.1 | - | 1, 2 | 200 | 40047070 | ||
| JQ080910.1 | - | 1, 2 | 200 | |||

Wfikkn2
Genomic Map