siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 9 | 687 | ||
| GenBank mRNA | AK029489.1 | - | 8 | 512 | ||
| AK037748.1 | - | 3 | 2618 | |||
| AK050534.1 | - | 9 | 697 | |||
| BC008086.1 | - | 9 | 687 | 3586161 | ||
| BC030675.1 | - | 9 | 685 | 1068780 | ||
| BC037025.1 | - | 5 | 2044 | |||
| BC083081.1 | - | 9 | 721 | 6510396 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108443.

Zfp330
Genomic Map