siRNA Details
| Gene Symbol: Celf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176954.3 | NP_795928.2 | 11 | 1192 | ||
| GenBank mRNA | AK029796.1 | - | 11 | 2366 | ||
| AK132321.1 | - | 11 | 1158 | |||
| AK158669.1 | - | 10 | 969 | |||
| BC079845.1 | - | 11 | 1163 | |||

Celf5
Genomic Map