siRNA Details
| Gene Symbol: Prps1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 1338 | ||
| GenBank mRNA | AK134500.1 | - | 2 | 1339 | ||
| BC110357.1 | - | 2 | 1241 | 5293637 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prps1l3| Gene Symbol: Prps1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 1338 | ||
| GenBank mRNA | AK134500.1 | - | 2 | 1339 | ||
| BC110357.1 | - | 2 | 1241 | 5293637 | ||