siRNA Details
| Gene Symbol: Olfr192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207549.1 | NP_997432.1 | 1 | 803 | ||
| Gene Symbol: Olfr190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146397.2 | NP_666509.2 | 1 | 803 | ||
| GenBank mRNA | BC104050.1 | - | 1 | 803 | 40040225 | |
| BC104051.1 | - | 1 | 803 | 40040226 | ||

Olfr192
Genomic Map