siRNA Details
| Gene Symbol: G630018N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090209.1 | - | 8 | 1006 | ||
| AK143085.1 | - | 5 | 4553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G630018N14Rik| Gene Symbol: G630018N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090209.1 | - | 8 | 1006 | ||
| AK143085.1 | - | 5 | 4553 | |||