siRNA Details
| Gene Symbol: Gm5464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034881.3 | NP_001030053.1 | 1 | 1357 | ||
| GenBank mRNA | AK135012.1 | - | 1 | 1358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5464| Gene Symbol: Gm5464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034881.3 | NP_001030053.1 | 1 | 1357 | ||
| GenBank mRNA | AK135012.1 | - | 1 | 1358 | ||