siRNA Details
| Gene Symbol: Gm5464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034881.3 | NP_001030053.1 | 1 | 997 | ||
| GenBank mRNA | AK135012.1 | - | 1 | 998 | ||
| BC147346.1 | - | 1 | 995 | 9056391 | ||
| BC147347.1 | - | 1 | 995 | 9056397 | ||

Gm5464
Genomic Map