siRNA Details
| Gene Symbol: Defb45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037752.2 | NP_001032841.1 | 2 | 200 | ||
| GenBank mRNA | AK005919.1 | - | 2 | 201 | ||
| AY591384.1 | - | 2 | 145 | |||
| BC132226.1 | - | 2 | 140 | 40130503 | ||
| BC132228.1 | - | 2 | 140 | 40130505 | ||
| DQ012038.1 | - | 2 | 197 | |||

Defb45
Genomic Map